Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azido-PEG5-acid | 1425973-16-5 | MFCD23726612 | ≥98.0% | 335.35 | C13H25N3O7 | 1 G
SDP

Supplier:  Medchemexpress LLC HY1305721G

Encompass_Preferred

Azido-PEG5-acid is a PEG-based, azide-terminated 5-unit linker used for bioconjugation and click chemistry. It is water soluble and commonly used as a linker in PROTAC and ADC synthesis, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC).

  • Azide functional group for CuAAC and SPAAC
  • Five-unit PEG spacer provides water solubility
  • Molecular weight 335.35 g/mol
  • CAS number 1425973-16-5
  • Purity ≥98.0%
  • Appearance: colorless to light yellow liquid
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)

Catalog No. 50-002-04288


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.