Learn More
Medchemexpress LLC Azido-PEG6-acid | 361189-66-4 | 99.77% | C15H29N3O8 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY140453500MG
Encompass_Preferred
Azido-PEG6-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group. This allows it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules possessing Alkyne groups. Additionally, it can engage in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an Azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Engages in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-52941