Learn More
Medchemexpress LLC Azido-PEG6-alcohol | 86770-69-6 | 98.0% | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY130537
Encompass_Preferred
Azido-PEG6-alcohol is a PEG-based PROTAC linker and a non-cleavable 6-unit PEG ADC linker. It is designed for the synthesis of PROTACs and antibody-drug conjugates (ADCs). This compound features an Azide group, making it suitable for click chemistry reactions, including copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules and strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules possessing DBCO or BCN groups.
  • Used in the synthesis of PROTACs
  • Acts as a non-cleavable 6-unit PEG ADC linker
  • Facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Utilizes an Azide group for click chemistry applications
Catalog No. 50-003-42706