Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azido-PEG6-C2-Boc | 406213-76-1 | 99.8% | C19H37N3O8 | 1 G
SDP

Supplier:  Medchemexpress LLC HY1407791G

Encompass_Preferred

Azido-PEG6-C2-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an azide group that can participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing alkyne groups. It also facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) reactions when combined with molecules containing DBCO or BCN groups.

  • Biochemical category
  • Regulatory status: RUO
  • Research field: Cancer
  • Solubility: 10 mM in DMSO
  • Grade: No development reported
  • Purity: 99.8%
  • Molecular formula: C19H37N3O8
  • Molecular weight: 435.51
  • CAS-Number: 406213-76-1
  • Storage conditions: -20°C for 3 years, 4°C for 2 years (powder)
  • Shipping at room temperature

Catalog No. 50-003-52966


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.