Learn More
Medchemexpress LLC Azido-PEG6-C2-Boc | 406213-76-1 | 99.8% | C19H37N3O8 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY140779500MG
Encompass_Preferred
Azido-PEG6-C2-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Azide group that can participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Click chemistry reagent with Azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Allows strain-promoted alkyne-azide cycloaddition (SPAAC)
Catalog No. 50-003-53361