Learn More
Medchemexpress LLC Azido-PEG6-MS | 352439-38-4 | 95.3% | 385.43 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY138363
Encompass_Preferred
Azido-PEG6-MS is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This compound functions as a click chemistry reagent, incorporating an azide group that enables it to participate in copper-catalyzed azide-alkyne cycloaddition reactions with molecules possessing alkyne groups. Additionally, it can engage in strain-promoted alkyne-azide cycloaddition reactions with molecules containing DBCO or BCN groups.
  • It is a PEG-based PROTAC linker.
  • It can be used in the synthesis of PROTACs.
  • It is a click chemistry reagent.
  • It contains an azide group.
  • It can undergo copper-catalyzed azide-alkyne cycloaddition reactions with molecules containing alkyne groups.
  • It can also undergo strain-promoted alkyne-azide cycloaddition reactions with molecules containing DBCO or BCN groups.
Catalog No. 50-003-50478