Learn More
Medchemexpress LLC Azido-PEG6-MS | 352439-38-4 | 95.3% | 385.43 | 50 MG

Supplier: Medchemexpress LLC HY13836350MG
Azido-PEG6-MS is a PEG-based PROTAC linker and click chemistry reagent. It contains an Azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions. PROTACs utilize intracellular ubiquitin-proteasome systems to selectively degrade target proteins.
- Purity: 95.32%
- CAS number: 352439-38-4
- Molecular weight: 385.43
- Chemical formula: C13H27N3O8S
- Appearance: Liquid
- Color: Colorless to light yellow
- Click chemistry: Azide group for CuAAc and SPAAC reactions
- PROTAC synthesis: Functions as a PROTAC linker
- Disease applications: Cancer and cancer targeted therapy
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.