Learn More
Medchemexpress LLC Azido-PEG6-MS | 352439-38-4 | 95.32% | 385.43 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY138363100MG
Encompass_Preferred
Azido-PEG6-MS is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, suitable for copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker for PROTAC synthesis.
  • Click chemistry reagent.
  • Contains an azide group.
  • Enables copper-catalyzed azide-alkyne cycloaddition reactions.
  • Enables strain-promoted alkyne-azide cycloaddition reactions.
Catalog No. 50-003-48625