Learn More
eMolecules​ Azido-PEG6-NHS ester | Broadpharm | 2055014-64-5 | MFCD22683276 | 476.483 | C19H32N4O10 | 95.000 | [N-]=[N+]=NCCOCCOCCOCCOCCOCCOCCC(=O)ON1C(=O)CCC1=O | 1g | 296213826
Supplier:  eMolecules​ 296213826
Encompass_Preferred
Azido-PEG6-NHS ester | Broadpharm | 2055014-64-5 | MFCD22683276 | 476.483 | C19H32N4O10 | 95.000 | [N-]=[N+]=NCCOCCOCCOCCOCCOCCOCCC(=O)ON1C(=O)CCC1=O | 1g | 296213826
Catalog No. NC1655533