Learn More
Medchemexpress LLC Azido-PEG6-NHS ester | 2055014-64-5 | MFCD22683276 | 98.0% | 476.48 g/mol | C19H32N4O10 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY13047425MG
Encompass_Preferred
Azido-PEG6-NHS ester is a bifunctional PEG-based reagent containing an azide group for bioorthogonal click chemistry and an N-hydroxysuccinimide (NHS) ester for coupling to primary amines. It is commonly used as a linker in antibody-drug conjugate and PROTAC synthesis, and for general bioconjugation applications.
  • Bifunctional: azide for CuAAC/SPAAC and NHS ester for amine coupling.
  • PEG6 spacer improves solubility and reduces steric hindrance.
  • High purity for reliable conjugation reactions.
  • Suitable for ADC and PROTAC linker installation and bioconjugation workflows.
  • Available in small-scale packaging compatible with research applications.
Catalog No. 50-002-03380