Learn More
Medchemexpress LLC Azido-PEG7-azide | 225523-86-4 | MFCD29764341 | >96.0% | 420.46 g/mol | C16H32N6O7 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1333945MG
Encompass_Preferred
Azido-PEG7-azide is a homobifunctional polyethylene glycol (PEG) linker bearing terminal azide groups used for click chemistry and bioconjugation. It provides a seven-unit ethylene glycol spacer that increases hydrophilicity and flexibility, facilitating conjugation in synthesis workflows. Molecular formula C16H32N6O7; molecular weight 420.46 g/mol; CAS 225523-86-4.
  • Contains two terminal azide groups for click chemistry.
  • Seven-unit PEG spacer increases hydrophilicity and solubility.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) reactions.
  • Serves as a flexible linker for PROTAC and other bioconjugation applications.
  • Supplied in small research pack sizes suitable for synthesis and analysis.
Catalog No. 50-002-62627