Learn More
Medchemexpress LLC Azido-PEG8-azide | 361543-07-9 | 98.2% | 464.51 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY138706100MG
Encompass_Preferred
Azido-PEG8-azide is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. This click chemistry reagent features an Azide group, enabling participation in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules that have DBCO or BCN groups.
  • Functions as a click chemistry reagent
  • Features an azide group
  • Compatible with CuAAc reactions
  • Compatible with SPAAC reactions
Catalog No. 50-003-50059