Learn More
Medchemexpress LLC Azido-PEG8-azide | 361543-07-9 | 98.2% | 464.51 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY138706500MG
Encompass_Preferred
Azido-PEG8-azide is a PEG-based PROTAC linker designed for the synthesis of PROTACs. This click chemistry reagent contains an Azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Alkyne groups. It also facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based linker for PROTAC synthesis.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc).
  • Supports strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Useful for synthesizing PROTACs to degrade target proteins.
Catalog No. 50-003-49695