Learn More
Medchemexpress LLC Azido-PEG8-CH2COO-PFP | 2182601-80-3 | 98.0% | C24H34F5N3O10 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14077450MG
Encompass_Preferred
Azido-PEG8-CH2COO-PFP is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is a click chemistry reagent containing an Azide group, which can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with Alkyne groups or strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
Catalog No. 50-003-55692