Learn More
Medchemexpress LLC Azido-PEG8-hydrazide | 2353410-11-2 | 96.1% | 481.54 g/mol | C19H39N5O9 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1408155MG
Encompass_Preferred
Azido-PEG8-hydrazide is an azide-terminated PEG8 linker with a hydrazide functional group, designed for use in click chemistry and as a versatile linker for assembling bifunctional molecules. It is provided as a solid for research applications and is compatible with common bioconjugation reactions.
Catalog No. 50-002-23241