Learn More
Medchemexpress LLC Azido-PEG8-hydrazide | 2353410-11-2 | 96.11% | C19H39N5O9 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14081550MG
Encompass_Preferred
Azido-PEG8-hydrazide is a PEG-based PROTAC linker designed for the synthesis of PROTACs. It functions as a click chemistry reagent, capable of participating in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Participates in CuAAc reactions
  • Engages in SPAAC reactions
  • Utilized in the synthesis of PROTACs
Catalog No. 50-003-56499