Learn More
Medchemexpress LLC Azido-PEG9-acid | 1670249-37-2 | 97.7% | 511.56 g/mol | C21H41N3O11 | 1 G

Supplier: Medchemexpress LLC HY1304751G
Azido-PEG9-acid is an azide-terminated 9-unit polyethylene glycol linker with a terminal carboxylic acid. It is used as a non-cleavable PEG linker for conjugation chemistry and click-based bioconjugation applications.
- Non-cleavable 9-unit PEG linker suitable for ADC and PROTAC synthesis.
- Azide functional group enables CuAAC and strain-promoted azide-alkyne cycloaddition (SPAAC).
- Molecular weight 511.56 g/mol; formula C21H41N3O11.
- Purity approximately 97.7%.
- Appearance: colorless to light yellow liquid; storage: -20°C for long-term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.