Learn More
Medchemexpress LLC Azido-PEG9-acid | 1670249-37-2 | 97.7% | 511.56 g/mol | C21H41N3O11 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13047550MG
Encompass_Preferred
Azido-PEG9-acid is an azido-functional polyethylene glycol linker containing nine ethylene glycol units, used as a hydrophilic spacer and azide handle for conjugation chemistry in small-molecule and bioconjugate synthesis. It is suitable for click chemistry, antibody-drug conjugate (ADC) linker assembly, and PROTAC construction.
  • Azido functional group for efficient click chemistry conjugation.
  • Nine-unit PEG spacer to increase solubility and reduce steric hindrance.
  • Non-cleavable linker suitable for ADC and PROTAC synthesis.
  • Molecular weight 511.56 g/mol and defined chemical formula.
  • Available in small-mass research quantities for synthetic applications.
Catalog No. 50-002-03911