Learn More
Medchemexpress LLC Azido-PEG9-amine | 1207714-69-9 | MFCD30723241 | 98.5% | 482.57 g/mol | C20H42N4O9 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY130169100MG
Encompass_Preferred
Azido-PEG9-amine is an azide-terminated nine-unit polyethylene glycol (PEG) linker with a terminal primary amine used for bioconjugation and linker design. It is suitable for click chemistry and linker-based constructs, providing hydrophilic spacing between functional groups.
  • Azide-terminated PEG linker compatible with CuAAC and SPAAC click reactions.
  • Contains a primary amine for orthogonal functionalization.
  • Nine ethylene glycol units provide hydrophilicity and spacing for conjugates.
  • Reported high purity supports conjugation and analytical workflows.
  • Available in small research-scale quantities for method development.
Catalog No. 50-002-03943