Learn More
Medchemexpress LLC Azido-PEG9-NHS ester | 2801772-87-0 | C25H44N4O13 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY140758
Encompass_Preferred
Azido-PEG9-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent with an azide group, capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAC) with alkyne-containing molecules. It also undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Target PROTAC linkers
  • Used in PROTAC pathway
  • Enables selective degradation of target proteins via the intracellular ubiquitin-proteasome system
Catalog No. 50-003-52664