Learn More
Apexbio Technology LLC Azidobutyric acid NHS ester 50mg
SDP
Supplier:  Apexbio Technology LLC A814750
Encompass_Preferred
Azidobutyric acid NHS ester is a chemical reagent used to introduce azide functional groups onto biomolecules containing primary amines It is designed to enable the covalent modification of proteins peptides or amino-modified nucleotides facilitating subsequent copper-catalyzed azide-alkyne click chemistry reactions Azidobutyric acid NHS ester exerts its utility by forming stable amide bonds with amino groups under mild aqueous conditions thus providing an azide handle for bioorthogonal labeling Based on these properties Azidobutyric acid NHS ester holds research potential in applications such as bioimaging flow cytometry protein profiling affinity purification and drug conjugation research
Catalog No. 50-000-92560