Learn More
Medchemexpress LLC Azilsartan mepixetil | 1596357-16-2 | MFCD34676898 | 98.1% | 678.69 g·mol⁻1 | C36H34N6O8 | 5 MG

Supplier: Medchemexpress LLC HY1455525MG
Azilsartan mepixetil is a prodrug angiotensin II receptor antagonist used in preclinical research. It is applied in studies of hypertension, chronic heart failure, and diabetic nephropathy and is reported to deliver potent, long-acting antihypertensive effects. The material is characterized for identity and purity to support laboratory research applications.
- Chemical name: Azilsartan mepixetil.
- CAS number: 1596357-16-2.
- Purity: 98.1%.
- Molecular formula: C36H34N6O8.
- Molecular weight: 678.69 g·mol⁻1.
- Formulation: solid powder; also available as 10 mM solutions in DMSO when specified.
- Intended use: preclinical research into antihypertensive and cardioprotective effects.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.