Learn More
Medchemexpress LLC Azimsulfuron | 120162-55-2 | 98.0% | 5 MG

Supplier: Medchemexpress LLC HY1210795MG
Azimsulfuron is a sulfonylurea herbicide supplied as an analytical research standard for laboratory use. It is used to control broad-leaved and sedge weeds in paddy fields and is suitable for analytical assays, mode-of-action studies, and method development.
- Purity 98.0%.
- Typical package sizes 5 MG, 10 MG, 25 MG, 50 MG, 100 MG.
- CAS number 120162-55-2.
- Molecular formula C13H16N10O5S.
- Molecular weight 424.40.
- Appearance white to off-white solid.
- Intended for research and analytical applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.