Learn More
Selleck Chemical LLC Azithromycin 25mg 83905-01-5 CP-62993, XZ-450
SDP
Supplier:  Selleck Chemical LLC S183525MG
Encompass_Preferred
Azithromycin (CP-62993, XZ-450) is an antibiotic by inhibiting protein synthesis, used for the treatment of bacterial infections. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1177379