Learn More
Medchemexpress LLC Azithromycin-d5 | 83905-01-5 | 754.02 | C38H67D5N2O12 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW6538751MG
Encompass_Preferred
Azithromycin-d5 is the deuterium-labeled analogue of azithromycin, a macrolide antibiotic. It is supplied for research use as an internal standard and tracer in quantitative analytical workflows, supporting pharmacokinetic and metabolic studies where isotopically labeled standards improve accuracy and precision.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for LC-MS, GC-MS, and NMR applications.
  • Offered in small-scale analytical packaging (e.g., 1 mg).
  • Molecular formula C38H67D5N2O12; molecular weight 754.02.
  • Certificate of analysis/data sheet available for analytical use.
Catalog No. 50-002-84240