Learn More
Medchemexpress LLC Azithromycin-d5 (CP-62993-d5) | 83905-01-5 | MFCD00873574 | >95.0% | 754.02 | C38H67D5N2O12 | 500 UG
SDP
Supplier:  Medchemexpress LLC HYW653875500UG
Encompass_Preferred
Azithromycin-d5 is the deuterium-labeled analogue of azithromycin supplied for research and analytical applications. It is intended as a stable-isotope internal standard or tracer for mass spectrometry, providing closely matched behavior to the unlabeled compound for accurate quantitation in LC-MS/MS assays.
  • Deuterium-labeled azithromycin (d5) for use as an internal standard.
  • Suitable for LC-MS/MS and quantitative analysis.
  • Molecular formula C38H67D5N2O12; molecular weight 754.02.
  • Available in small, research-scale amounts for analytical workflows.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-84959