Learn More
Medchemexpress LLC Azithromycin hydrate | 117772-70-0 | 99.4% | 785.02 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY17506A50MG
Encompass_Preferred
Azithromycin hydrate is a macrolide antibiotic useful for the treatment of a number of bacterial infections.
  • Useful for the treatment of various bacterial infections.
  • Augments rhinovirus-induced IFNβ expression in primary bronchial epithelial cells from asthmatics.
  • Associated with over-expression of RIG-I like receptors and repression of viral replication.
  • Specifically reduces MMP-9 messenger RNA and protein levels.
  • Augments expression of interferon-stimulated genes and the pattern recognition receptor MDA5.
Catalog No. 50-003-87040