Learn More
Sapidyne Instruments Inc Azlactone Activated Beads, Pack of 10 vials, suitable for coupling small molecules (<20 kDa), covalent immobilization via amide bonds
SDP
Supplier:  Sapidyne Instruments Inc 444110
Encompass
Azlactone Activated Beads, Pack of 10 vials, suitable for coupling small molecules (<20 kDa), covalent immobilization via amide bonds
Catalog No. NC0353450