Learn More
Medchemexpress LLC HY-B0529A 500mg Medchemexpress, Azlocillin (sodium salt) CAS:37091-65-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0529A500MG
Encompass_Preferred
Medchemexpress, HY-B0529A 500mg Azlocillin (sodium salt) CAS:37091-65-9 Azlocillin sodium salt (Sodium azlocillin), a semisynthetic penicillin, is a broad spectrum β-lactam antibiotic. Azlocillin sodium salt shows antipseudomonal activity, and also potent against the malarial parasite Plasmodium falciparum. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2583