Learn More
Medchemexpress LLC HY-B2127 1g Medchemexpress, Azobenzene CAS:103-33-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB21271G
Encompass_Preferred
Medchemexpress, HY-B2127 1g Azobenzene CAS:103-33-3 Azobenzene can be used as an optical trigger for the design and synthesis of a large variety of photoresponsive systems. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3073