Learn More
Medchemexpress LLC Azoreductase/NQO1 H 500ug
SDP
Supplier:  Medchemexpress LLC HYP74405A500UG
Encompass_Preferred
Azo reductase/NQO1 protein is a flavin-containing quinone reductase that uses NADH or NADPH to catalyze the two-electron reduction of quinone to hydroquinone It regulates cellular redox status by detoxifying quinones and reducing plasma membrane redox components Azoreductase/NQO1 Protein Human is the recombinant human-derived Azoreductase/NQO1 protein expressed by E coli with tag free
Catalog No. 50-001-78815