Learn More
Medchemexpress LLC HY-B0849 50mg Medchemexpress, Azoxystrobin CAS:131860-33-8 Purity:>98%
Supplier: Medchemexpress LLC HYB084950MG
Medchemexpress, HY-B0849 50mg Azoxystrobin CAS:131860-33-8 Azoxystrobin is a broad-spectrum β-methoxyacrylate fungicide. Azoxystrobin inhibits mitochondrial respiration by binding to the Qo site of the cytochrome bc1 complex and inhibiting electron transfer. Azoxystrobin induces the production of reactive oxygen species (ROS) and induces cell apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.