Learn More
Medchemexpress LLC Aztreonam (Standard) | 78110-38-0 | 72145 | 99.6% | 25 MG
Supplier: Medchemexpress LLC 25MGHYB0129R
Aztreonam (Standard) is an analytical standard and synthetic monocyclic beta-lactam antibiotic. It is intended for research and analytical applications, serving as a reference standard for assays.
- Used in qualitative, quantitative, and methodological research experiments
- Utilized with techniques such as HPLC, GC, and MS
- Possesses a very high affinity for penicillin-binding protein 3 (PBP-3)