Learn More
Electron Microscopy Sciences Azur A Certified 10 GR

Supplier: Electron Microscopy Sciences 11240
(Methylene Azur A)
CAS #531-53-3 C₁₄H₁₄ClN₃S F.W. 291.80
Solubility: Water-soluble; Alcohol-soluble
Water and alcohol-soluble. Used in nuclear staining; A polychromatic blood stain.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.