Learn More
Medchemexpress LLC HY-D0004 5mg Medchemexpress, Azure B CAS:531-55-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYD00045MG
Encompass_Preferred
Medchemexpress, HY-D0004 5mg Azure B CAS:531-55-5 Azure B is a cationic dye and the major metabolite of Methylene blue. Azure B is used in making Azure eosin stains for blood smear staining. Azure B is a high-potency, selective and reversible inhibitor of monoamine oxidases (MAO)-A, with IC50s of 11 and 968 nM for recombinant human MAO-A and MAO-B, respectively. Azure B possesses significant antidepressant-like effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0182