Learn More
Medchemexpress LLC Azurocidin Protein, Human (HEK293, His) | ≥98.0% | Approximately 35-44 kDa due to glycosylation | 250 UG

Supplier: Medchemexpress LLC 250UGHYP7624
Azurocidin Protein, Human (HEK293, His) is a recombinant human Azurocidin produced in HEK293 cells with a His tag. It is a protein rapidly mobilized from emigrating polymorphonuclear leukocytes (PMN) and acts as an important mediator during the initiation of the immune response. It functions as a chemoattractant and activator of monocytes and macrophages, enhancing cytokine release and bacterial phagocytosis, and also activates endothelial cells, causing vascular leakage and edema formation.
- Recombinant human Azurocidin
- Produced in HEK293 cells
- Features a His tag
- Protein length: partial
- Glycosylation: yes
- Endotoxin level: <1 EU/μg
- Appearance: lyophilized powder
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.