Learn More
Medchemexpress LLC HY-19314A 5mg Medchemexpress, Azvudine (hydrochloride) CAS:1333126-31-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY19314A5MG
Encompass_Preferred
Medchemexpress, HY-19314A 5mg Azvudine (hydrochloride) CAS:1333126-31-0 Azvudine (RO-0622) hydrochloride is a potent nucleoside reverse transcriptase inhibitor (NRTI), with antiviral activity on HIV, HBV and HCV. Azvudine hydrochloride exerts highly potent inhibition on HIV-1 (EC50s ranging from 0.03 to 6.92 nM) and HIV-2 (EC50s ranging from 0.018 to 0.025 nM). Azvudine hydrochloride inhibits NRTI-resistant viral strains. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0845