Learn More
ABclonal Technology -Actin Rabbit mAb 100 L
SDP
Supplier:  ABclonal Technology AC038
Encompass_Preferred
Immunogen: Recombinant protein of human β-Actin•Host Species: Rabbit•Isotype: IgG•Species Reactivity:Human,Mouse,Rat,Chicken,Zebrafish,Pig•Validated Applications:WB•Swiss Prot: P60709•Gene ID:60•Purification Method:Affinity purification•Storage: Store at -20℃. Avoid freeze / thaw cycles.|Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.This product is available in 20UL, 50UL, 100UL, and 200UL. Please contact customer service for pricing and availability on alternative sizes.
Catalog No. NC2079925