Learn More
Medchemexpress LLC B-AMINOPROPIONITRILE
SDP
Supplier:  Medchemexpress LLC HYY175010MM 1ML IN DMSO
Encompass_Preferred
β-Aminopropionitrile is a specific and irreversible lysyl oxidase (LOX) inhibitor. β-Aminopropionitrile targets the active site of LOX or LOXL isoenzymes.
Catalog No. NC1840354