Learn More
AnaSpec B AMYLOIED (1-40) HILYTE FLUR
Catalog No. NC0501493
Encompass
Beta-Amyloid (1-40), HiLyte(TM) Fluor 488-labeled, Human[HiLyte(TM) Fluor 488-DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVV] ; MW 4686.3; (0.1 mg) This is a fluorescent (HiLyte(TM) Fluor 488)-labeled B-Amyloid peptide, Abs/Em=503/528 nm.
Catalog No. NC0501493 Supplier AnaSpec Supplier No. AS6049101
Beta-Amyloid (1-40), HiLyte(TM) Fluor 488-labeled, Human[HiLyte(TM) Fluor 488-DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVV] ; MW 4686.3; (0.1 mg) This is a fluorescent (HiLyte(TM) Fluor 488)-labeled B-Amyloid peptide, Abs/Em=503/528 nm.