Learn More
Selleck Chemical LLC b-AP15 10mg 1009817-63-3 NSC687852
SDP
Supplier:  Selleck Chemical LLC S492010MG
Encompass_Preferred
b-AP15 (NSC687852) is a deubiquitinases inhibitor for 19S proteasomes activity of Ub-AMC cleavage with IC50 of 2.1 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4299