Learn More
Ubpbio b-AP15, 2MG
Supplier:  Ubpbio F2100
Encompass_Preferred
b-AP15 is a specific inhibitor of the deubiquitinating enzymes Uch37 and Usp14 of the 26S proteasome. It blocks the deubiquitinating activity of the 26S proteasome. Treating cells with b-AP15 causes the accumulation of polyubiquitinated proteins, inhibition of proteasomal degradation, upregulation of apoptotic markers and cell death.
Catalog No. 50-196-5101