Learn More
Medchemexpress LLC B-catenin-IN-37 10mg | 1783856-40-5 | 307.27 g·mol⁻¹ | C13H9N9O | 10 MG

Supplier: Medchemexpress LLC HY11554310MG
β-catenin-IN-37 is a small-molecule inhibitor of the β-catenin/T-cell factor protein-protein interaction that suppresses canonical Wnt signaling and reduces proliferation of Wnt-activated colorectal cancer cell lines in vitro.
- Selective β-catenin/Tcf PPI inhibitor.
- Reported IC50 values: 20 μM (SW480) and 31 μM (HCT116).
- Molecular formula C13H9N9O; molecular weight 307.27 g·mol⁻¹.
- High reported purity (99.13%).
- Supplied as a 10 mg research reagent for in vitro assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.