Learn More
Enzo Life Sciences beta-Crystallin (bovine), (native) (1mg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2241000
Encompass_Preferred
Alternative name: CRYBB. Purity: ≥80% (SDS-PAGE; Western blot). Formulation: Liquid. In PBS, pH 7.2, containing 0.15M sodium chloride, 0.05M phosphate buffer, and 0.09% sodium azide. Source: Isolated from bovine eye lens. Long Term Storage: -80°C. Applications: WB .
Catalog No. 50-201-4022