Learn More
Medchemexpress LLC B-D-glucopyranosiduronic acid derivative, Fmoc-protected methyl ester, 2,3,4-triacetate | 894096-02-7 | 98.8% | 748.73 g·mol^-1 | C38H40N2O14 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13108750MG
Encompass_Preferred
Me-triacetyl-β-D-glucopyranuronate-Ph-CH2OH-Fmoc is a glycosylated, cleavable linker intermediate used in the synthesis of antibody-drug conjugates (ADCs). It provides a protected glucuronic acid moiety with an Fmoc-protected aminopropionyl spacer for selective conjugation and stability during synthetic workflows.
  • Cleavable glycosidic linker for ADC synthesis.
  • High purity (assay 98.8%).
  • Molecular formula C38H40N2O14, molecular weight 748.73 g·mol^-1.
  • Available in small-scale quantities suitable for synthesis (for example, 50 MG).
  • Supplied with certificate of analysis for batch verification.
Catalog No. 50-002-02375