Learn More
Alpco Ltd Free Beta Human Chorionic Gonadotropin (HCG) ELISA
SDP
Supplier:  Alpco Ltd 20FBEHUE01
Encompass_Preferred
The Free Beta Human Chorionic Gonadotropin (? HCG) ELISA is an enzyme immunoassay for the quantitative determination of the free beta subunit of human chorionic gonadotropin (free ? hCG) in human serum and EDTA plasma. For research use only. Not for use in diagnostic procedures.
Catalog No. NC2885431