Learn More
TARGETMOL CHEMICALS INC B I09 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1484710MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. B I09 an IRE-1 RNase inhibitor with an IC50 of 1230 nM inhibits splicing of XBP1 mRNA in human WaC3 cells and expression of xbp-1 in LPS-stimulated B cells. B I09 can be used to simulate the defects of XBP-1 in CLL cells. an IRE-1 RNase inhibitor with an IC50 of 1230 nM inhibits splicing of XBP1 mRNA in human WaC3 cells and expression of xbp-1 in LPS-stimulated B cells. B I09 can be used to simulate the defects of XBP-1 in CLL cells. an IRE-1 RNase inhibitor with an IC50 of 1230 nM inhibits splicing of XBP1 mRNA in human WaC3 cells and expression of xbp-1 in LPS-stimulated B cells. B I09 can be used to simulate the defects of XBP-1 in CLL cells. purity: 99%
Catalog No. 50-258-1279