Learn More
ATCC ATCC Quantitative Genomic RNA, Influenza B (Yamagata lineage) strain B/Massachusetts/2/2012 - 100 μL, ≥1x10^5 copies/μL (VR-1813DQ)
Supplier:  ATCC VR1813DQ
Encompass
Quantitative Genomic RNA from Influenza B virus (Yamagata Lineage) strain B/Massachusetts/2/2012 supplied as a quantified RNA standard for molecular assay development, verification, and validation. The quantitative format supports generation of qPCR standard curves to determine viral load and to monitor assay performance.
  • Quantitative genomic RNA from Influenza B (Yamagata lineage)
  • Suitable for qPCR standard curves and viral load determination
  • Concentration ≥1 x 10^5 copies/μL, volume 100 μL
  • Stored at -70°C or colder
  • Intended for assay development, verification, and validation
Catalog No. NC3760737