Learn More
Medchemexpress LLC HY-F0004 50mg , β-Nicotinamide mononucleotide CAS:1094-61-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYF000450MG
Encompass_Preferred
HY-F0004 50mg β-Nicotinamide mononucleotide CAS: 1094-61-7 β-nicotinamide mononucleotide (β-NM) is a product of the nicotinamide phosphoribosyltransferase (NAMPT) reaction and a key NAD+ intermediate. The pharmacological activities of β-nicotinamide mononucleotide include its role in cellular biochemical functions, cardioprotection, diabetes, Alzheimer's disease, and complications associated with obesity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9211