Learn More
Medchemexpress LLC B-Raf IN 1 | 950736-05-7 | 99.9% | 515.53 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1822710MM/1ML
Encompass_Preferred
B-Raf IN 1 is a potent and selective B-Raf kinase inhibitor with an IC50 of 24 nM. It is for research use only and appears as a light yellow to yellow solid.
  • Potent and selective B-Raf kinase inhibitor
  • IC50 of 24 nM for B-Raf
  • Appears as a solid
  • Light yellow to yellow color
  • For research use only
Catalog No. 50-003-87585